accessibility menu, dialog, popup

UserName

Viewing profile as user:
OPOOFSn

CAS RN 52262-58-5

IUPAC Name: λ²-tin(2+) phosphorofluoridate
Synonyms: λ²-tin(2+) fluorophosphate
Molecular Weight (g/mol): 216.68
Molecular Formula: FO3PSn
InChi Key: ZIQLCYCKKROENE-UHFFFAOYSA-L
SMILES: [Sn++].[O-]P([O-])(F)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results