Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tin(II) fluorophosphate, Thermo Scientific Chemicals

Catalog No. AA3960806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3960806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3960806 Supplier Thermo Scientific Chemicals Supplier No. 03960806

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52262-58-5
Molecular Formula FO3PSn
Molecular Weight (g/mol) 216.679
MDL Number MFCD00058794
InChI Key ZIQLCYCKKROENE-UHFFFAOYSA-L
Synonym tin ii fluorophosphate, stannous fluorophosphate, acmc-20ajhz, fluoridophosphoric acid tin ii salt, tin, ion sn2+ fluoro-phosphite ion, fluoro-dioxido-oxophosphorane; tin 2+, fluoranyl-bis oxidanidyl-oxidanylidene-$l^ 5-phosphane; tin 2+
PubChem CID 44717639
IUPAC Name fluoro-dioxido-oxo-$l^{5}-phosphane;tin(2+)
SMILES [O-]P(=O)([O-])F.[Sn+2]

Specifications

Color White
Physical Form Powder
Quantity 5 g
Solubility Information Slightly soluble (3.0g/L at 25°C)
Formula Weight 216.66
Chemical Name or Material Tin(II) fluorophosphate
Hazard Category H302-H314-H335
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.