CAS RN 5231-87-8
3,4-Diethoxy-3-cyclobutene-1,2-dione 98.0+%, TCI America™
CAS: 5231-87-8 Molecular Formula: C8H10O4 Molecular Weight (g/mol): 170.164 InChI Key: DFSFLZCLKYZYRD-UHFFFAOYSA-N Synonym: 3,4-diethoxy-3-cyclobutene-1,2-dione,diethyl squarate,squaric acid diethyl ester,3-cyclobutene-1,2-dione, 3,4-diethoxy,diethoxycyclobutenedione,diethoxycyclobut-3-ene-1,2-dione,1,2-diethoxycyclobutenedione,3,4-diethoxy-cyclobut-3-ene-1,2-dione,pubchem9741,acmc-209kyn PubChem CID: 123228 IUPAC Name: 3,4-diethoxycyclobut-3-ene-1,2-dione SMILES: CCOC1=C(C(=O)C1=O)OCC
3,4-Diethoxy-3-cyclobutene-1,2-dione, 98%
CAS: 5231-87-8 Molecular Formula: C8H10O4 Molecular Weight (g/mol): 170.16 MDL Number: MFCD00001333 InChI Key: DFSFLZCLKYZYRD-UHFFFAOYSA-N Synonym: 3,4-diethoxy-3-cyclobutene-1,2-dione,diethyl squarate,squaric acid diethyl ester,3-cyclobutene-1,2-dione, 3,4-diethoxy,diethoxycyclobutenedione,diethoxycyclobut-3-ene-1,2-dione,1,2-diethoxycyclobutenedione,3,4-diethoxy-cyclobut-3-ene-1,2-dione,pubchem9741,acmc-209kyn PubChem CID: 123228 IUPAC Name: 3,4-diethoxycyclobut-3-ene-1,2-dione SMILES: CCOC1=C(C(=O)C1=O)OCC
3,4-Diethoxy-3-cyclobutene-1,2-dione, 98%
CAS: 5231-87-8 Molecular Formula: C8H10O4 Molecular Weight (g/mol): 170.164 MDL Number: MFCD00001333 InChI Key: DFSFLZCLKYZYRD-UHFFFAOYSA-N Synonym: 3,4-diethoxy-3-cyclobutene-1,2-dione,diethyl squarate,squaric acid diethyl ester,3-cyclobutene-1,2-dione, 3,4-diethoxy,diethoxycyclobutenedione,diethoxycyclobut-3-ene-1,2-dione,1,2-diethoxycyclobutenedione,3,4-diethoxy-cyclobut-3-ene-1,2-dione,pubchem9741,acmc-209kyn PubChem CID: 123228 IUPAC Name: 3,4-diethoxycyclobut-3-ene-1,2-dione SMILES: CCOC1=C(C(=O)C1=O)OCC
3,4-diethoxycyclobut-3-ene-1,2-dione, Thermo Scientific™
CAS: 5231-87-8 Molecular Formula: C8H10O4 Molecular Weight (g/mol): 170.164 InChI Key: DFSFLZCLKYZYRD-UHFFFAOYSA-N Synonym: 3,4-diethoxy-3-cyclobutene-1,2-dione,diethyl squarate,squaric acid diethyl ester,3-cyclobutene-1,2-dione, 3,4-diethoxy,diethoxycyclobutenedione,diethoxycyclobut-3-ene-1,2-dione,1,2-diethoxycyclobutenedione,3,4-diethoxy-cyclobut-3-ene-1,2-dione,pubchem9741,acmc-209kyn PubChem CID: 123228 IUPAC Name: 3,4-diethoxycyclobut-3-ene-1,2-dione SMILES: CCOC1=C(C(=O)C1=O)OCC
Sigma Aldrich 3,4-Diethoxy-3-cyclobutene-1,2-dione
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.