accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3HOCH3CH3HOCH3CH3HOCH3CH3HOCH3CH3HOCH3CH3HOW

CAS RN 52321-90-1

IUPAC Name: hexakis(propan-2-ol) tungsten
Synonyms: hexakis(isopropyl alcohol) tungsten
Molecular Weight (g/mol): 544.42
Molecular Formula: C18H48O6W
InChi Key: ZGWZVFZMNDLXGR-UHFFFAOYSA-N
SMILES: [W].CC(C)O.CC(C)O.CC(C)O.CC(C)O.CC(C)O.CC(C)O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results