Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tungsten(VI) isopropoxide, 5% w/v in isopropanol, Thermo Scientific™

Catalog No. AA40247AE
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40247AE 100 mL
AA40247AC 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA40247AE Supplier Thermo Scientific Chemicals Supplier No. 40247AE

Precursor for Tungsten oxide using sol-gel process

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52321-90-1
Molecular Formula C18H48O6W
Molecular Weight (g/mol) 544.416
MDL Number MFCD00210652
InChI Key ZGWZVFZMNDLXGR-UHFFFAOYSA-N
Synonym tungsten vi isopropoxide, tungsten vi isopropoxide w/v in isopropanol, tungsten vi isopropoxide w/v in isopropanol, packaged under argon in resealable chemseal bottles
PubChem CID 121233707
IUPAC Name propan-2-ol;tungsten
SMILES CC(C)O.CC(C)O.CC(C)O.CC(C)O.CC(C)O.CC(C)O.[W]

Specifications

Linear Formula W[OCH(CH3)2]6
Quantity 100 mL
UN Number UN1219
Sensitivity Moisture Sensitive
Formula Weight 538.38
Concentration or Composition (by Analyte or Components) 5% w/v in isopropanol
Physical Form Liquid
Chemical Name or Material Tungsten(VI) isopropoxide
Hazard Category Danger
Hazard Statement GHS H Statement
H225-H314-H336
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Precautionary Statement GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.