accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 52346-13-1

IUPAC Name: tert-butyl 2-(1-benzoyl-3-ethenylpiperidin-4-yl)acetate
Synonyms: tert-butyl 2-(1-benzoyl-3-ethenylpiperidin-4-yl)acetate
Molecular Weight (g/mol): 329.44
Molecular Formula: C20H27NO3
InChi Key: LYFYOJAKMXFONP-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)CC1CCN(CC1C=C)C(=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results