Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzoylmeroquinene tert-Butyl Ester 98.0+%, TCI America™
SDP

Catalog No. B23611G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B23611G Supplier TCI America Supplier No. B23611G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 52346-13-1
Molecular Formula C20H27NO3
Molecular Weight (g/mol) 329.44
MDL Number MFCD08276275
InChI Key LYFYOJAKMXFONP-UHFFFAOYNA-N
Synonym (3R,4S)-1-Benzoyl-3-vinyl-4-piperidineacetic Acid tert-Butyl Ester, tert-Butyl (3R,4S)-1-Benzoyl-3-vinyl-4-piperidineacetate
PubChem CID 11809651
IUPAC Name tert-butyl 2-(1-benzoyl-3-ethenylpiperidin-4-yl)acetate
SMILES CC(C)(C)OC(=O)CC1CCN(CC1C=C)C(=O)C1=CC=CC=C1

Specifications

Melting Point 64°C
Color White-Yellow
Quantity 1 g
Formula Weight 329.44
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N-Benzoylmeroquinene tert-Butyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.