accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5238-22-2

H3CCH3OOOOCH3CH3

CAS RN 5238-22-2

IUPAC Name: 1,8-bis(2-ethylhexyl) octanedioate
Synonyms: 1,8-bis(2-ethylhexyl) octanedioate
Molecular Weight (g/mol): 398.63
Molecular Formula: C24H46O4
InChi Key: FTALILOVQKMLBH-UHFFFAOYNA-N
SMILES: CCCCC(CC)COC(=O)CCCCCCC(=O)OCC(CC)CCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)