Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-ethylhexyl) Suberate 98.0+%, TCI America™
SDP

Catalog No. S044325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S044325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S044325ML Supplier TCI America Supplier No. S044325ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5238-22-2
Molecular Formula C24H46O4
Molecular Weight (g/mol) 398.628
MDL Number MFCD01631311
InChI Key FTALILOVQKMLBH-UHFFFAOYSA-N
Synonym Dioctyl Suberate, Suberic Acid Dioctyl Ester, Suberic Acid Bis(2-ethylhexyl) Ester
PubChem CID 6452627
IUPAC Name bis(2-ethylhexyl) octanedioate
SMILES CCCCC(CC)COC(=O)CCCCCCC(=O)OCC(CC)CCCC

Specifications

Color Colorless
Quantity 25 mL
Formula Weight 398.63
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(2-ethylhexyl) Suberate
EINECSNumber (2)-2569
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.