accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 52387-15-2

IUPAC Name: (1S)-1-(5,8-dihydroxy-1,4-dioxo-1,4-dihydronaphthalen-2-yl)-4-methylpent-3-en-1-yl (2S)-2-methylbutanoate
Synonyms: (1S)-1-(5,8-dihydroxy-1,4-dioxonaphthalen-2-yl)-4-methylpent-3-en-1-yl (2S)-2-methylbutanoate
Molecular Weight (g/mol): 372.42
Molecular Formula: C21H24O6
InChi Key: ODQATBANLZCROD-SJCJKPOMSA-N
SMILES: CC[C@H](C)C(=O)O[C@@H](CC=C(C)C)C1=CC(=O)C2=C(O)C=CC(O)=C2C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results