Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Methylbutyryl)shikonin 88.0+%, TCI America™
SDP

Catalog No. M1028100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1028100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1028100MG Supplier TCI America Supplier No. M1028100MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 52387-15-2
Molecular Formula C21H24O6
Molecular Weight (g/mol) 372.42
MDL Number MFCD00142946
InChI Key ODQATBANLZCROD-SJCJKPOMSA-N
Synonym 5,8-Dihydroxy-2-[1-(2-methylbutyryloxy)-4-methyl-3-pentenyl]-1,4-naphthalenedione
PubChem CID 10429346
IUPAC Name (1S)-1-(5,8-dihydroxy-1,4-dioxo-1,4-dihydronaphthalen-2-yl)-4-methylpent-3-en-1-yl (2S)-2-methylbutanoate
SMILES CC[C@H](C)C(=O)O[C@@H](CC=C(C)C)C1=CC(=O)C2=C(O)C=CC(O)=C2C1=O

Specifications

Color Red
Quantity 100 mg
Formula Weight 372.42
Percent Purity ≥88.0% (HPLC)
Physical Form Paste
Chemical Name or Material (2-Methylbutyryl)shikonin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.