accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5240-72-2

IUPAC Name: {bicyclo[2.2.1]heptan-2-yl}methanol
Synonyms: 2-norbornane methanol
Molecular Weight (g/mol): 126.2
Molecular Formula: C8H14O
InChi Key: LWHKUVOYICRGGR-UHFFFAOYNA-N
SMILES: OCC1CC2CCC1C2
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results