Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Norbornane-2-methanol 95.0+%, TCI America™
SDP

Catalog No. N055325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N055325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N055325ML Supplier TCI America Supplier No. N055325ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5240-72-2
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00074722
InChI Key LWHKUVOYICRGGR-UHFFFAOYSA-N
Synonym 2-Hydroxymethylbicyclo[2.2.1]heptane
PubChem CID 21308
IUPAC Name 3-bicyclo[2.2.1]heptanylmethanol
SMILES C1CC2CC1CC2CO

Specifications

Boiling Point 108°C
Quantity 25 mL
UN Number 2810
Formula Weight 126.20
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Norbornane-2-methanol
RTECSNumber RB7350000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.