accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NHCl

CAS RN 52417-22-8

IUPAC Name: acridin-9-amine hydrochloride
Synonyms: aminacrine hydrochloride
Molecular Weight (g/mol): 230.7
Molecular Formula: C13H11ClN2
InChi Key: FTGPOQQGJVJDCT-UHFFFAOYSA-N
SMILES: Cl.NC1=C2C=CC=CC2=NC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (4)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (5)
  • (4)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)