Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Aminoacridine Hydrochloride Monohydrate 99+%, Thermo Scientific™

Catalog No. AC103110250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC103110250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC103110250 Supplier Thermo Scientific Chemicals Supplier No. 103110250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52417-22-8 , 134-50-9
Molecular Formula C13H11ClN2
Molecular Weight (g/mol) 230.70
MDL Number MFCD00012663
InChI Key FTGPOQQGJVJDCT-UHFFFAOYSA-N
Synonym 9-aminoacridine hydrochloride monohydrate, acridin-9-amine hydrochloride hydrate, 9-aminoacridine hydrochloride hydrate, 9-acridinamine, monohydrochloride, monohydrate, 9-aminoacridine, monohydrochloride, monohydrate, acridin-9-ylamine, dsstox_cid_4457, dsstox_rid_77405, dsstox_gsid_24457, acridine-9-ylamine, chloride, hydrate
PubChem CID 2723598
IUPAC Name acridin-9-amine hydrochloride
SMILES Cl.NC1=C2C=CC=CC2=NC2=CC=CC=C12

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color Yellow
Loss on Drying 8% max. (105°C)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Quantity 25 g
Beilstein 22, 463
Merck Index 14, 407
Formula Weight 248.71
Percent Purity 99.0 to 101.0% (on dried substance)
Physical Form Fine Crystalline Powder
Chemical Name or Material 9-Aminoacridine hydrochloride monohydrate, 99+%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Toxic if inhaled.
Toxic in contact with skin.
Suspected of causing genetic defects.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.