accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 52431-30-8

IUPAC Name: 2,5-dibromo-3,4-dinitrothiophene
Synonyms: 2,5-dibromo-3,4-dinitrothiophene
Molecular Weight (g/mol): 331.92
Molecular Formula: C4Br2N2O4S
InChi Key: AHGHPBPARMANQD-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(Br)SC(Br)=C1[N+]([O-])=O
Quantity 
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)