Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dibromo-3,4-dinitrothiophene, Thermo Scientific Chemicals

Catalog No. AC278220050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC278220050 5 g Glass bottle
AC278220010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC278220050 Supplier Thermo Scientific Chemicals Supplier No. 278220050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 52431-30-8 | C4Br2N2O4S | 331.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52431-30-8
Molecular Formula C4Br2N2O4S
Molecular Weight (g/mol) 331.92
MDL Number MFCD00015537
InChI Key AHGHPBPARMANQD-UHFFFAOYSA-N
Synonym 2,5-dibromo-3,4-dinitro-thiophene, thiophene, 2,5-dibromo-3,4-dinitro, acmc-1apgk, ksc491i5j, 2,5-dibromo-3,4-dinitrothiophene gc
PubChem CID 257024
IUPAC Name 2,5-dibromo-3,4-dinitrothiophene
SMILES [O-][N+](=O)C1=C(Br)SC(Br)=C1[N+]([O-])=O

Specifications

Melting Point 132.0°C to 134.0°C
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 331.94
Chemical Name or Material 2, 5-Dibromo-3, 4-dinitrothiophene
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.