accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH3C

CAS RN 52480-43-0

IUPAC Name: 4,5-dimethylfuran-2-carbaldehyde
Synonyms: 4,5-dimethylfuran-2-carbaldehyde
Molecular Weight (g/mol): 124.14
Molecular Formula: C7H8O2
InChi Key: JPTPEPVCVXGNJM-UHFFFAOYSA-N
SMILES: CC1=C(C)C=C(O1)C=O
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results