Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Dimethylfurfural 97.0+%, TCI America™
SDP

Catalog No. D26391G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D26391G Supplier TCI America Supplier No. D26391G

Specifications

CAS 52480-43-0
Boiling Point 100°C
Molecular Formula C7H8O2
Quantity 1 g
MDL Number MFCD00085030
Synonym 4,5-dimethyl-2-furaldehyde, 4,5-dimethylfurfural, 4,5-dimethyl-furan-2-carbaldehyde, 2-furancarboxaldehyde, 4,5-dimethyl, 2-furanmethanethiolformate, acmc-1ao31, 4.5-dimethyl-2-furaldehyde, 4,5-dimethyl-2-furancarbaldehyde, 4,5-dimethyl-2-furancarboxaldehyde, 2-furancarboxaldehyde,4,5-dimethyl
InChI Key JPTPEPVCVXGNJM-UHFFFAOYSA-N
SMILES CC1=C(OC(=C1)C=O)C
IUPAC Name 4,5-dimethylfuran-2-carbaldehyde
Molecular Weight (g/mol) 124.139
PubChem CID 643406
Formula Weight 124.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4,5-Dimethylfurfural
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.