accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 52536-09-1

IUPAC Name: sodium pyridin-3-olate
Synonyms: sodium pyridin-3-olate
Molecular Weight (g/mol): 117.08
Molecular Formula: C5H4NNaO
InChi Key: WHYQZPOBOBSAIS-UHFFFAOYSA-M
SMILES: [Na+].[O-]C1=CC=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results