Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxypyridine Sodium Salt 98%, Thermo Scientific™

Catalog No. AC134610500
Change view
Click to view available options
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC134610500 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC134610500 Supplier Thermo Scientific Chemicals Supplier No. 134610500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 52536-09-1
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C5H4NNaO
Linear Formula C5H4NONa
Quantity 50g
Synonym sodium pyridin-3-olate, 3-hydroxypyridine sodium salt, 3-sodiooxy pyridine, sodium 3-pyridinolate, sodium pyridine-3-olate, 3-hydroxypyridine sodium salt?
InChI Key WHYQZPOBOBSAIS-UHFFFAOYSA-M
SMILES C1=CC(=CN=C1)[O-].[Na+]
IUPAC Name sodium;pyridin-3-olate
Molecular Weight (g/mol) 117.083
PubChem CID 2723769
Formula Weight 117.08
Percent Purity ≥97.5%
Chemical Name or Material 3-Hydroxypyridine sodium salt, 98%
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 257-995-1
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.