accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(R)HOO(R)(S)(S)(S)OH(R)H3C(S)(R)H3C

CAS RN 5255-17-4

IUPAC Name: (4R)-4-[(1R,3aS,3bS,7S,9aR,9bS,11aR)-7-hydroxy-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]pentanoic acid
Synonyms: cholenic acid 3β-hydroxy-5-cholenic acid D5-cholenate 3b-hydroxy-5-cholenoate 3b-hydroxychol-5-enoate 3β-hydroxy-5-cholenoic acid 3b-hydroxycholenoate 3b-hydroxy-5-cholenoic acid 3b-hydroxychol-5-enoic acid 3b-hydroxycholenoic acid
Molecular Weight (g/mol): 374.57
Molecular Formula: C24H38O3
InChi Key: HIAJCGFYHIANNA-QIZZZRFXSA-N
SMILES: C[C@H](CCC(O)=O)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results