Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3beta-Hydroxy-DELTA5-cholenic Acid, TCI America™
SDP

Catalog No. H05215G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H05215G 5 g
H05211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H05215G Supplier TCI America Supplier No. H05215G

Chemical Identifiers

CAS 5255-17-4
Molecular Formula C24H38O3
Molecular Weight (g/mol) 374.565
MDL Number MFCD00069468
InChI Key HIAJCGFYHIANNA-QIZZZRFXSA-N
PubChem CID 92997
IUPAC Name (4R)-4-[(3S,8S,9S,10R,13R,14S,17R)-3-hydroxy-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
SMILES CC(CCC(=O)O)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C

Specifications

Melting Point 232°C
Color White-Yellow
Quantity 5 g
Formula Weight 374.57
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3beta-Hydroxy-DELTA5-cholenic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.