accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH2H2N

CAS RN 52562-19-3

IUPAC Name: 2-(prop-1-en-2-yl)aniline
Synonyms: 2-(prop-1-en-2-yl)aniline
Molecular Weight (g/mol): 133.19
Molecular Formula: C9H11N
InChi Key: HEDYZFYQYPWWCC-UHFFFAOYSA-N
SMILES: CC(=C)C1=CC=CC=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results