Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Isopropenylaniline, 98+%, Thermo Scientific™

Catalog No. AC186490050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC186490050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC186490050 Supplier Thermo Scientific Chemicals Supplier No. 186490050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52562-19-3
Molecular Formula C9H11N
Molecular Weight (g/mol) 133.194
MDL Number MFCD00007719
InChI Key HEDYZFYQYPWWCC-UHFFFAOYSA-N
Synonym 2-prop-1-en-2-yl aniline, 2-isopropenylaniline, o-isopropenylaniline, benzenamine, 2-1-methylethenyl
PubChem CID 96494
IUPAC Name 2-prop-1-en-2-ylaniline
SMILES CC(=C)C1=CC=CC=C1N

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Yellow
Density 0.97g/mL
Infrared Spectrum Authentic
Assay Percent Range 98+%
Refractive Index 1.5690 to 1.5710 (20°C, 589nm)
Linear Formula H2C=C(CH3)C6H4NH2
Quantity 5 g
Beilstein 12, III, 2793
Formula Weight 133.19
Percent Purity ≥98.0%
Physical Form Liquid
Chemical Name or Material 2-Isopropenylaniline, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.