accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 52605-52-4

IUPAC Name: 1-(3-chlorophenyl)-4-(3-chloropropyl)piperazin-1-ium chloride
Synonyms: 1-(3-chlorophenyl)-4-(3-chloropropyl)piperazin-1-ium chloride
Molecular Weight (g/mol): 309.66
Molecular Formula: C13H19Cl3N2
InChi Key: JVLRNANYLVRULL-UHFFFAOYSA-N
SMILES: [Cl-].ClCCCN1CC[NH+](CC1)C1=CC=CC(Cl)=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results