Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3-Chlorophenyl)-4-(3-chloropropyl)piperazine Hydrochloride 97.0+%, TCI America™
SDP

Catalog No. C160825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C160825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C160825G Supplier TCI America Supplier No. C160825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 52605-52-4
Molecular Formula C13H19Cl3N2
Molecular Weight (g/mol) 309.66
MDL Number MFCD00067039
InChI Key JVLRNANYLVRULL-UHFFFAOYSA-N
PubChem CID 13035120
IUPAC Name 1-(3-chlorophenyl)-4-(3-chloropropyl)piperazin-1-ium chloride
SMILES [Cl-].ClCCCN1CC[NH+](CC1)C1=CC=CC(Cl)=C1

Specifications

Melting Point 201°C
Color White-Yellow
Quantity 25 g
Formula Weight 309.66
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(3-Chlorophenyl)-4-(3-chloropropyl)piperazine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.