CAS RN 52646-27-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
D-(+)-Celloheptaose, 95%, Thermo Scientific Chemicals
CAS: 52646-27-2 MDL Number: MFCD15145329 Synonym: Celloheptaose