accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSOOCl

CAS RN 52665-49-3

IUPAC Name: furan-3-sulfonyl chloride
Synonyms: furan-3-sulfonyl chloride
Molecular Weight (g/mol): 166.58
Molecular Formula: C4H3ClO3S
InChi Key: PCBBEIXTVWWTAK-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=COC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results