Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Furan-3-sulfonyl chloride, ≥97%, Thermo Scientific™

Catalog No. CC02103DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC02103DA 1 g Amber Glass Bottle
CC02103CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC02103DA Supplier Thermo Scientific Chemicals Supplier No. CC02103DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 52665-49-3
Molecular Formula C4H3ClO3S
Molecular Weight (g/mol) 166.575
MDL Number MFCD08271875
InChI Key PCBBEIXTVWWTAK-UHFFFAOYSA-N
Synonym 3-furansulfonyl chloride, furan-3-sulfonylchloride, chloro-3-furylsulfone, 3-furansulfonylchlorid, 3-furansulfonylchloride, 3-furylsulfonyl chloride, 3-chlorosulphonyl furan, 3-furansulfonyl chloride;, 3-furanylsulfonyl chloride, 3-furylsulfonyl chloride;
PubChem CID 22179302
IUPAC Name furan-3-sulfonyl chloride
SMILES C1=COC=C1S(=O)(=O)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 166.58
Percent Purity 97%
Chemical Name or Material Furan-3-sulfonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Reacts violently with water.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.