accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5267-64-1

IUPAC Name: 2-amino-3-phenylpropan-1-ol
Synonyms: 2-amino-3-phenyl-1-propanol
Molecular Weight (g/mol): 151.21
Molecular Formula: C9H13NO
InChi Key: STVVMTBJNDTZBF-UHFFFAOYNA-N
SMILES: NC(CO)CC1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)