Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Phenylalaninol 98.0+%, TCI America™
SDP

Catalog No. P12895G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12895G 5 g
P128925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12895G Supplier TCI America Supplier No. P12895G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5267-64-1
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.209
MDL Number MFCD00064298
InChI Key STVVMTBJNDTZBF-SECBINFHSA-N
Synonym d-phenylalaninol, r-2-amino-3-phenylpropan-1-ol, d-penylalaninol, r-+-2-amino-3-phenyl-1-propanol, 2r-2-amino-3-phenylpropan-1-ol, d +-phenylalaninol, +-d-phenylalaninol, r-2-amino-3-phenyl-1-propanol, d-+-phenylalaninol, r-phenylalaninol
PubChem CID 853475
IUPAC Name (2R)-2-amino-3-phenylpropan-1-ol
SMILES C1=CC=C(C=C1)CC(CO)N

Specifications

Melting Point 93°C
Boiling Point 122°C
Quantity 5 g
UN Number 3259
Formula Weight 151.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material D-Phenylalaninol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.