CAS RN 527-07-1
Gluconic acid, sodium salt, 98%
CAS: 527-07-1 Molecular Formula: C6H11NaO7 Molecular Weight (g/mol): 218.137 MDL Number: MFCD00064210 InChI Key: UPMFZISCCZSDND-JJKGCWMISA-M Synonym: sodium gluconate,sodium d-gluconate,d-gluconic acid sodium salt,d-gluconic acid, monosodium salt,gluconic acid sodium salt,monosodium gluconate,glonsen,monosodium d-gluconate,gluconate sodium,pasexon 100t PubChem CID: 23672301 ChEBI: CHEBI:84997 IUPAC Name: sodium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate SMILES: C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Na+]
Sodium Gluconate, USP, 98-102%, Spectrum™ Chemical
CAS: 527-07-1 Molecular Formula: C6H11NaO7 Molecular Weight (g/mol): 218.14 InChI Key: UPMFZISCCZSDND-UHFFFAOYNA-M IUPAC Name: sodium 2,3,4,5,6-pentahydroxyhexanoate SMILES: [Na+].OCC(O)C(O)C(O)C(O)C([O-])=O
Sodium Gluconate, FCC, 98-102%, Spectrum™ Chemical
CAS: 527-07-1 Molecular Formula: C6H11NaO7 Molecular Weight (g/mol): 218.14 InChI Key: UPMFZISCCZSDND-UHFFFAOYNA-M IUPAC Name: sodium 2,3,4,5,6-pentahydroxyhexanoate SMILES: [Na+].OCC(O)C(O)C(O)C(O)C([O-])=O
D-Gluconic Acid sodium salt, Certified Reference Material, MilliporeSigma™ Supelco™
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Sodium Gluconate, FCC, 98-102%, Spectrum™ Chemical
CAS: 527-07-1 Molecular Formula: C6H11NaO7 Molecular Weight (g/mol): 218.14 InChI Key: UPMFZISCCZSDND-UHFFFAOYNA-M IUPAC Name: sodium 2,3,4,5,6-pentahydroxyhexanoate SMILES: [Na+].OCC(O)C(O)C(O)C(O)C([O-])=O
Sodium D-gluconate, 97%
CAS: 527-07-1 Molecular Formula: C6H11NaO7 Molecular Weight (g/mol): 218.137 MDL Number: MFCD00064210 InChI Key: UPMFZISCCZSDND-JJKGCWMISA-M Synonym: sodium gluconate,sodium d-gluconate,d-gluconic acid sodium salt,d-gluconic acid, monosodium salt,gluconic acid sodium salt,monosodium gluconate,glonsen,monosodium d-gluconate,gluconate sodium,pasexon 100t PubChem CID: 23672301 ChEBI: CHEBI:84997 IUPAC Name: sodium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate SMILES: C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Na+]
Sodium Gluconate 99.0+%, TCI America™
CAS: 527-07-1 Molecular Formula: C6H11NaO7 Molecular Weight (g/mol): 218.137 MDL Number: MFCD00064210 InChI Key: UPMFZISCCZSDND-JJKGCWMISA-M Synonym: sodium gluconate,sodium d-gluconate,d-gluconic acid sodium salt,d-gluconic acid, monosodium salt,gluconic acid sodium salt,monosodium gluconate,glonsen,monosodium d-gluconate,gluconate sodium,pasexon 100t PubChem CID: 23672301 ChEBI: CHEBI:84997 IUPAC Name: sodium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate SMILES: C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Na+]
Sodium Gluconate, USP, 98-102%, Spectrum™ Chemical
CAS: 527-07-1 Molecular Formula: C6H11NaO7 Molecular Weight (g/mol): 218.14 InChI Key: UPMFZISCCZSDND-UHFFFAOYNA-M IUPAC Name: sodium 2,3,4,5,6-pentahydroxyhexanoate SMILES: [Na+].OCC(O)C(O)C(O)C(O)C([O-])=O