Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Gluconate, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18606512 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-512 2.5 kg Poly Bottle
18-606-511 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-606-512 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S12862.5KG

missing translation for 'validMainframeMsg'

S1286, 527-07-1, C6H11NaO7

Spectrum™ Chemical Sodium Gluconate, USP is used as surface cleaning agent for metals and glass bottles. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 527-07-1
Molecular Formula C6H11NaO7
Molecular Weight (g/mol) 218.14
InChI Key UPMFZISCCZSDND-UHFFFAOYNA-M
IUPAC Name sodium 2,3,4,5,6-pentahydroxyhexanoate
SMILES [Na+].OCC(O)C(O)C(O)C(O)C([O-])=O

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 2.5 kg
Percent Purity 98 to 102%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.