accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 527-17-3

IUPAC Name: tetramethylcyclohexa-2,5-diene-1,4-dione
Synonyms: duroquinone
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: WAMKWBHYPYBEJY-UHFFFAOYSA-N
SMILES: CC1=C(C)C(=O)C(C)=C(C)C1=O
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)