accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5272-18-4

CH3SiH3COH

CAS RN 5272-18-4

IUPAC Name: dimethyl(phenyl)silanol
Synonyms: dimethyl(phenyl)silanol
Molecular Weight (g/mol): 152.27
Molecular Formula: C8H12OSi
InChi Key: FDTBETCIPGWBHK-UHFFFAOYSA-N
SMILES: C[Si](C)(O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results