Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylphenylsilanol, 97%, Thermo Scientific™

Catalog No. AAH2737703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2737703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2737703 Supplier Thermo Scientific Chemicals Supplier No. H2737703

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5272-18-4
Molecular Formula C8H12OSi
Molecular Weight (g/mol) 152.27
MDL Number MFCD08276879
InChI Key FDTBETCIPGWBHK-UHFFFAOYSA-N
PubChem CID 78929
IUPAC Name hydroxy-dimethyl-phenylsilane
SMILES C[Si](C)(O)C1=CC=CC=C1

Specifications

Boiling Point 60°C to 62°C (0.5mmHg)
Quantity 1 g
Synonym dimethylphenylsilanol, phenyldimethylsilanol, silanol, dimethylphenyl, silanol, 1,1-dimethyl-1-phenyl, dimethyl phenyl silanol, acmc-1avoz, amtsi002, silanol,1,1-dimethyl-1-phenyl
Formula Weight 152.27
Percent Purity 97%
Chemical Name or Material Dimethylphenylsilanol
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.