accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaH3CHNNOSOOOH3C(Z)

CAS RN 52749-23-2

IUPAC Name: sodium (5Z)-5-[2-(2,3-dimethylphenyl)hydrazin-1-ylidene]-6-oxo-5,6-dihydronaphthalene-2-sulfonate
Synonyms: sodium (5Z)-5-[2-(2,3-dimethylphenyl)hydrazin-1-ylidene]-6-oxonaphthalene-2-sulfonate
Molecular Weight (g/mol): 378.38
Molecular Formula: C18H15N2NaO4S
InChi Key: SEVBKEFOLSVXMD-GQQUDWLYSA-M
SMILES: [Na+].CC1=CC=CC(N\N=C2/C(=O)C=CC3=CC(=CC=C23)S([O-])(=O)=O)=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results