Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Orange 17, TCI America™
SDP

Catalog No. B077825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B077825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B077825G Supplier TCI America Supplier No. B077825G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 52749-23-2
Molecular Formula C18H15N2NaO4S
Molecular Weight (g/mol) 378.38
MDL Number MFCD00059907
InChI Key SEVBKEFOLSVXMD-GQQUDWLYSA-M
Synonym Brilliant Orange H
PubChem CID 131706962
IUPAC Name sodium (5Z)-5-[2-(2,3-dimethylphenyl)hydrazin-1-ylidene]-6-oxo-5,6-dihydronaphthalene-2-sulfonate
SMILES [Na+].CC1=CC=CC(N\N=C2/C(=O)C=CC3=CC(=CC=C23)S([O-])(=O)=O)=C1C

Specifications

Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 378.38
Chemical Name or Material Acid Orange 17
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.