accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHSN

CAS RN 52767-84-7

IUPAC Name: 5-propyl-1,2-dihydropyrimidine-2-thione
Synonyms: 5-propyl-1H-pyrimidine-2-thione
Molecular Weight (g/mol): 154.23
Molecular Formula: C7H10N2S
InChi Key: YZFGTZRDMRKBBU-UHFFFAOYSA-N
SMILES: CCCC1=CNC(=S)N=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results