accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONONOO

CAS RN 528-29-0

IUPAC Name: 1,2-dinitrobenzene
Synonyms: O-dinitrobenzene
Molecular Weight (g/mol): 168.11
Molecular Formula: C6H4N2O4
InChi Key: IZUKQUVSCNEFMJ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1[N+]([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results