Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,2-Dinitrobenzene

Catalog No. NC3453285
Encompass
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NC3453285 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NC3453285 Supplier Sigma Aldrich Supplier No. 1266325G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,2-Dinitrobenzene
TRUSTED_SUSTAINABILITY

Specifications

CAS 528-29-0
Melting Point 114°C to 117°C (lit.)
Boiling Point 319°C (773 mmHg)
Molecular Formula C6H4N2O4
Linear Formula C6H4(NO2)2
MDL Number MFCD00007093
Quantity 500 mg
Molecular Weight (g/mol) 168.11
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.