accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3COHO

CAS RN 528-90-5

IUPAC Name: 2,4,5-trimethylbenzoic acid
Synonyms: benzoic acid, 2,4,5-trimethyl-
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: QENJZWZWAWWESF-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C=C1C)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results