accessibility menu, dialog, popup

UserName

Viewing profile as user:
ON

CAS RN 52805-36-4

IUPAC Name: 4-(benzyloxy)benzonitrile
Synonyms: 4-(benzyloxy)benzonitrile
Molecular Weight (g/mol): 209.25
Molecular Formula: C14H11NO
InChi Key: UDAOJHAASAWVIQ-UHFFFAOYSA-N
SMILES: N#CC1=CC=C(OCC2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results