Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzyloxybenzonitrile 98.0+%, TCI America™
SDP

Catalog No. B13655G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13655G 5 g
B136525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B13655G Supplier TCI America Supplier No. B13655G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 52805-36-4
Molecular Formula C14H11NO
Molecular Weight (g/mol) 209.248
MDL Number MFCD00079701
InChI Key UDAOJHAASAWVIQ-UHFFFAOYSA-N
Synonym 4-benzyloxybenzonitrile, 4-benzyloxy benzonitrile, 4-benzyloxy-benzonitrile, benzonitrile, 4-phenylmethoxy, 4-phenylmethoxy benzenecarbonitrile, maybridge1_005982, benzonitrile analogue, 5a, acmc-209l1v, 4-cyanophenoxy phenyl methane
PubChem CID 561371
IUPAC Name 4-phenylmethoxybenzonitrile
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)C#N

Specifications

Melting Point 94°C
Color White
Quantity 5 g
Formula Weight 209.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyloxybenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.