accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONBrOH

CAS RN 52805-45-5

IUPAC Name: 3-bromo-4-hydroxy-5-methoxybenzonitrile
Synonyms: 3-bromo-4-hydroxy-5-methoxybenzonitrile
Molecular Weight (g/mol): 228.05
Molecular Formula: C8H6BrNO2
InChi Key: FHXLXRNDUJYBEQ-UHFFFAOYSA-N
SMILES: COC1=C(O)C(Br)=CC(=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results