Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-4-hydroxy-5-methoxybenzonitrile 98.0+%, TCI America™
SDP

Catalog No. B47575G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47575G 5 g
B47571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47575G Supplier TCI America Supplier No. B47575G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 52805-45-5
Molecular Formula C8H6BrNO2
Molecular Weight (g/mol) 228.045
MDL Number MFCD01123027
InChI Key FHXLXRNDUJYBEQ-UHFFFAOYSA-N
Synonym 3-bromo-4-hydroxy-5-methoxy-benzonitrile, 3-bromo-4-hydroxy-5-methoxybenzenecarbonitrile, acmc-1atrh, 3-bromo-4-hydroxy-5-methoxybenzonitrile
PubChem CID 7021625
IUPAC Name 3-bromo-4-hydroxy-5-methoxybenzonitrile
SMILES COC1=C(C(=CC(=C1)C#N)Br)O

Specifications

Melting Point 146°C
Color White
Quantity 5 g
UN Number 3439
Formula Weight 228.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-4-hydroxy-5-methoxybenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.