accessibility menu, dialog, popup

UserName

Viewing profile as user:
CaCH3HNNOOOSOOO(Z)

CAS RN 5281-04-9

IUPAC Name: calcium (4Z)-4-[2-(4-methyl-2-sulfonatophenyl)hydrazin-1-ylidene]-3-oxo-3,4-dihydronaphthalene-2-carboxylate
Synonyms: pigment red 57:1
Molecular Weight (g/mol): 424.44
Molecular Formula: C18H12CaN2O6S
InChi Key: WANIKCLUTOABTP-LRZQPWDCSA-L
SMILES: [Ca++].CC1=CC=C(N\N=C2/C(=O)C(=CC3=CC=CC=C23)C([O-])=O)C(=C1)S([O-])(=O)=O
Percent Purity 
  • (1)
  • (1)
Color 
  • (1)
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results