Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithol Rubin BCA 85.0+%, TCI America™
SDP

Catalog No. L007525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L007525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L007525G Supplier TCI America Supplier No. L007525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5281-04-9
Molecular Formula C18H12CaN2O6S
Molecular Weight (g/mol) 424.44
MDL Number MFCD00059529
InChI Key WANIKCLUTOABTP-LRZQPWDCSA-L
Synonym Brilliant Carmine 6B, Pigment Red 57
PubChem CID 121233252
IUPAC Name calcium;(4Z)-4-[(4-methyl-2-sulfonatophenyl)hydrazinylidene]-3-oxonaphthalene-2-carboxylate
SMILES CC1=CC(=C(C=C1)NN=C2C3=CC=CC=C3C=C(C2=O)C(=O)[O-])S(=O)(=O)[O-].[Ca].[Ca]

Specifications

Color Red
Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 424.44
Percent Purity ≥85.0% (E)
Chemical Name or Material Lithol Rubin BCA
EINECSNumber (5)-3244
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.