accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClSOOCl

CAS RN 52819-14-4

IUPAC Name: 3,4-dichloro-1λ⁶-thiophene-1,1-dione
Synonyms: 3,4-dichloro-1λ⁶-thiophene-1,1-dione
Molecular Weight (g/mol): 185.02
Molecular Formula: C4H2Cl2O2S
InChi Key: CIDAIMONSYOOGS-UHFFFAOYSA-N
SMILES: ClC1=CS(=O)(=O)C=C1Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results